Product Details

SNP ID
rs9357557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:47699447 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGGACCAGAGGCAATTGCTGGGC[C/G]TATAAATTTCTTTAAAATTTTATAA
Phenotype
MIM: 614268
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADGRF2 PubMed Links

Gene Details

Gene
ADGRF2
Gene Name
adhesion G protein-coupled receptor F2
There are no transcripts associated with this gene.

Gene
ADGRF4
Gene Name
adhesion G protein-coupled receptor F4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153838.3 Intron NP_722580.3

View Full Product Details