Product Details

SNP ID
rs6057110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:10035559 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAAATGTTTTACAAGGATCATAT[C/G]TCATTTCCTTTTCACACAGTCCCAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKEF1 PubMed Links

Gene Details

Gene
ANKEF1
Gene Name
ankyrin repeat and EF-hand domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303472.1 Intron NP_001290401.1
NM_022096.5 Intron NP_071379.3
NM_198798.2 Intron NP_942093.1
XM_005260792.2 Intron XP_005260849.1
Gene
SNAP25-AS1
Gene Name
SNAP25 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details