Product Details

SNP ID
rs10221509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4049552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATCCCTGCCCTCTCGAGGCTGGC[A/G]GCGGCCGAGGTGTTTGGTGACAGAT
Phenotype
MIM: 605878
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB7A PubMed Links

Gene Details

Gene
ZBTB7A
Gene Name
zinc finger and BTB domain containing 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317990.1 Intron NP_001304919.1
NM_015898.3 Intron NP_056982.1
XM_005259570.4 Intron XP_005259627.3
XM_005259571.4 Intron XP_005259628.1

View Full Product Details