Product Details

SNP ID
rs10225824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:117371969 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAATTAGTCTTAATGAATGAAAC[A/G]AAAATATATACCTTTCTTGTGACAT
Phenotype
MIM: 605797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASZ1 PubMed Links

Gene Details

Gene
ASZ1
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301821.1 Intron NP_001288750.1
NM_001301822.1 Intron NP_001288751.1
NM_130768.2 Intron NP_570124.1

View Full Product Details