Product Details

SNP ID
rs9481043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110191903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTAGATGATATATAATTTCTTTAC[A/G]ATTTTTAAAATAGAACATGAGTGCA
Phenotype
MIM: 605585 MIM: 605035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC40 PubMed Links

Gene Details

Gene
CDC40
Gene Name
cell division cycle 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015891.2 Intron NP_056975.1
Gene
WASF1
Gene Name
WAS protein family member 1
There are no transcripts associated with this gene.

View Full Product Details