Product Details
- SNP ID
-
rs6128728
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:59965376 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTTATGTGGGTTATATTTGCTGA[C/T]ACCATGATAGCTATTAAAACAGGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH26
PubMed Links
Gene Details
- Gene
- CDH26
- Gene Name
- cadherin 26
There are no transcripts associated with this gene.
View Full Product Details