Product Details

SNP ID
rs6955055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4778665 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATATGCTGAAGGGAATTTAATC[A/G]GGATGAGGGGTTCCACCTTTCTGGG
Phenotype
MIM: 613653 MIM: 616302
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP5Z1 PubMed Links

Gene Details

Gene
AP5Z1
Gene Name
adaptor related protein complex 5 zeta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014855.2 Intron NP_055670.1
XM_017012864.1 Intron XP_016868353.1
Gene
FOXK1
Gene Name
forkhead box K1
There are no transcripts associated with this gene.

Gene
MIR4656
Gene Name
microRNA 4656
There are no transcripts associated with this gene.

View Full Product Details