Product Details

SNP ID
rs7517517
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:4662891 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTATAAAGGGGAATATTTTGAAG[C/T]ATTAGAAGAAGTCCTATCTGGTCAG
Phenotype
MIM: 610972
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AJAP1 PubMed Links

Gene Details

Gene
AJAP1
Gene Name
adherens junctions associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042478.1 Intron NP_001035943.1
NM_018836.3 Intron NP_061324.1
XM_011541786.2 Intron XP_011540088.1
XM_011541787.2 Intron XP_011540089.1

View Full Product Details