Product Details

SNP ID
rs7530964
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:20186159 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATGGAGCGGAACGGCTAGGTGAG[A/C]GCGGCTGCCCGATCCCGCGGCCTGC
Phenotype
MIM: 616783
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G2C PubMed Links

Gene Details

Gene
PLA2G2C
Gene Name
phospholipase A2 group IIC
There are no transcripts associated with this gene.

Gene
UBXN10
Gene Name
UBX domain protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152376.4 Intron NP_689589.1
XM_005245742.3 Intron XP_005245799.1
XM_011540699.2 Intron XP_011539001.1
Gene
UBXN10-AS1
Gene Name
UBXN10 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details