Product Details

SNP ID
rs10251574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23183721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCCAGAAACAAATGAATGTTT[C/T]TGGAATTATAAGTATGAAGACCAAA
Phenotype
MIM: 611119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL7 PubMed Links

Gene Details

Gene
KLHL7
Gene Name
kelch like family member 7
There are no transcripts associated with this gene.

Gene
NUPL2
Gene Name
nucleoporin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007342.2 Intron NP_031368.1
XM_005249592.1 Intron XP_005249649.1
XM_005249593.3 Intron XP_005249650.1
XM_011515104.2 Intron XP_011513406.1
XM_017011714.1 Intron XP_016867203.1
XM_017011715.1 Intron XP_016867204.1
XM_017011716.1 Intron XP_016867205.1
XM_017011717.1 Intron XP_016867206.1
XM_017011718.1 Intron XP_016867207.1

View Full Product Details