Product Details

SNP ID
rs7996046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95439833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAAATAATAATAGTAAATAAATA[C/T]CAAACAACAATAATGAATAAATGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN10 PubMed Links

Gene Details

Gene
CLDN10
Gene Name
claudin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160100.1 Intron NP_001153572.1
NM_006984.4 Intron NP_008915.1
NM_182848.3 Intron NP_878268.1
XM_011521134.2 Intron XP_011519436.1
XM_017020843.1 Intron XP_016876332.1
XM_017020844.1 Intron XP_016876333.1

View Full Product Details