Product Details

SNP ID
rs10426369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47714507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGGACATCACTACAGATCCTA[C/T]GGTGAATAAAAGGCTAGTAAGGGAA
Phenotype
MIM: 605890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EHD2 PubMed Links

Gene Details

Gene
EHD2
Gene Name
EH domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014601.3 Intron NP_055416.2

View Full Product Details