Product Details

SNP ID
rs10129377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55057315 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCTGGAAATACCAGAGAATCAAG[C/T]GAGAAACTTTGAAGAATAAAATAAT
Phenotype
MIM: 616337
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPK1IP1L PubMed Links

Gene Details

Gene
MAPK1IP1L
Gene Name
mitogen-activated protein kinase 1 interacting protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144578.3 Intron NP_653179.1
XM_011537362.2 Intron XP_011535664.2
XM_017021789.1 Intron XP_016877278.1
Gene
SOCS4
Gene Name
suppressor of cytokine signaling 4
There are no transcripts associated with this gene.

View Full Product Details