Product Details
- SNP ID
-
rs9900684
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:29405924 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATTAAAATAGGTTCAGTAATGACA[C/T]ATCTGTATTTATTAATACATTTTTT
- Phenotype
-
MIM: 610266
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TAOK1
PubMed Links
Gene Details
- Gene
- TAOK1
- Gene Name
- TAO kinase 1
View Full Product Details