Product Details

SNP ID
rs9917722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:164982253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTACATTACTTACCATGTTGGTA[C/G]TGTCTTCATTAAAAGGAAGCGAATT
Phenotype
MIM: 609845
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SI PubMed Links

Gene Details

Gene
SI
Gene Name
sucrase-isomaltase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001041.3 5472 Missense Mutation ACT,AGT T1802S NP_001032.2
XM_011513078.2 5472 Missense Mutation ACT,AGT T1769S XP_011511380.1

View Full Product Details