Product Details

SNP ID
rs6624228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:66595017 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTTTATAATTCTACCAGCAATA[C/T]ATGAGGCTTCTCATTTCTCCACATC
Phenotype
MIM: 300276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EDA2R PubMed Links

Gene Details

Gene
EDA2R
Gene Name
ectodysplasin A2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199687.2 4152 Intron NP_001186616.1
NM_001242310.1 4152 Intron NP_001229239.1
NM_001324199.1 4152 Intron NP_001311128.1
NM_001324201.1 4152 Intron NP_001311130.1
NM_001324202.1 4152 Intron NP_001311131.1
NM_001324204.1 4152 Intron NP_001311133.1
NM_001324205.1 4152 Intron NP_001311134.1
NM_001324206.1 4152 Intron NP_001311135.1
NM_021783.4 4152 Intron NP_068555.1
XM_011531000.1 4152 UTR 3 XP_011529302.1
XM_011531001.1 4152 UTR 3 XP_011529303.1
XM_011531002.1 4152 UTR 3 XP_011529304.1
XM_011531003.2 4152 UTR 3 XP_011529305.1
XM_011531004.1 4152 Intron XP_011529306.1
XM_011531005.2 4152 UTR 3 XP_011529307.1
XM_011531006.2 4152 UTR 3 XP_011529308.1
XM_011531007.2 4152 Intron XP_011529309.1
XM_017029703.1 4152 UTR 3 XP_016885192.1
XM_017029704.1 4152 UTR 3 XP_016885193.1
XM_017029705.1 4152 UTR 3 XP_016885194.1
XM_017029706.1 4152 UTR 3 XP_016885195.1
XM_017029707.1 4152 UTR 3 XP_016885196.1
XM_017029708.1 4152 UTR 3 XP_016885197.1

View Full Product Details