Product Details
- SNP ID
-
rs9914865
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41634727 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGGGGTAGGGTGACCCTCCATC[C/T]TCAGCCACAGTTAGGAGGCTGAGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT42P
PubMed Links
Gene Details
- Gene
- KRT42P
- Gene Name
- keratin 42 pseudogene
There are no transcripts associated with this gene.
View Full Product Details