Product Details

SNP ID
rs6554164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:54240528 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCCCACCCAGGGGAAATCTTGGT[C/T]TTGCCTTTTCTTGTTTTCTACTCTA
Phenotype
MIM: 173490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDGFRA PubMed Links

Gene Details

Gene
PDGFRA
Gene Name
platelet derived growth factor receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006206.4 Intron NP_006197.1
XM_005265743.1 Intron XP_005265800.1
XM_006714039.3 Intron XP_006714102.1
XM_006714041.3 Intron XP_006714104.1
XM_011534385.2 Intron XP_011532687.1
XM_017008280.1 Intron XP_016863769.1
XM_017008281.1 Intron XP_016863770.1
XM_017008282.1 Intron XP_016863771.1

View Full Product Details