Product Details
- SNP ID
-
rs9982410
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:26837557 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTCCTCACTTTGCCTTGCCCTCA[A/G]AGCTAACACCACTTAAACCACTTAA
- Phenotype
-
MIM: 605174
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAMTS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116406019] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAMTS1
- Gene Name
- ADAM metallopeptidase with thrombospondin type 1 motif 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006988.4 |
3381 |
UTR 3 |
|
|
NP_008919.3 |
View Full Product Details