Product Details

SNP ID
rs6634764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130010829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCAGGAGCTTGGGGTGAGCCT[C/T]TAAAGAAGGGAGGCAAAAATTAGCC
Phenotype
MIM: 300688
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCORL1 PubMed Links

Gene Details

Gene
BCORL1
Gene Name
BCL6 corepressor-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184772.2 Intron NP_001171701.1
NM_021946.4 Intron NP_068765.3
XM_005262452.4 Intron XP_005262509.2
XM_005262453.4 Intron XP_005262510.2
XM_005262454.3 Intron XP_005262511.2
XM_005262455.4 Intron XP_005262512.2
XM_005262456.4 Intron XP_005262513.2
XM_006724776.3 Intron XP_006724839.1
XM_006724777.3 Intron XP_006724840.1
XM_006724779.2 Intron XP_006724842.1
XM_017029721.1 Intron XP_016885210.1
XM_017029722.1 Intron XP_016885211.1

View Full Product Details