Product Details

SNP ID
rs7519860
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:113939898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACATCCTTTCCTCATATTTTTCT[A/G]CTAACTTTCCTCGGTTCTTCAGAAT
Phenotype
MIM: 608003
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIPK1 PubMed Links

Gene Details

Gene
HIPK1
Gene Name
homeodomain interacting protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152696.3 Intron NP_689909.2
NM_181358.2 Intron NP_852003.1
NM_198268.2 Intron NP_938009.1
NM_198269.2 Intron NP_938010.1
XM_005270609.3 Intron XP_005270666.1
XM_005270610.4 Intron XP_005270667.1
XM_005270611.4 Intron XP_005270668.1
XM_005270612.4 Intron XP_005270669.1
XM_005270613.4 Intron XP_005270670.1
XM_006710443.3 Intron XP_006710506.1
XM_011540977.2 Intron XP_011539279.1
XM_017000605.1 Intron XP_016856094.1
XM_017000606.1 Intron XP_016856095.1

View Full Product Details