Product Details

SNP ID
rs7260223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56541283 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATGATTGGCTCATTTCTTGCCTT[C/T]TCCTTCCATATTTGGTCCTGATGCA
Phenotype
MIM: 616798
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP28 PubMed Links

Gene Details

Gene
ZFP28
Gene Name
ZFP28 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308440.1 Intron NP_001295369.1
NM_020828.1 Intron NP_065879.1
XM_011526462.2 Intron XP_011524764.1
XM_011526463.2 Intron XP_011524765.2

View Full Product Details