Product Details

SNP ID
rs10415806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41443569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGACAGAGGTGGGAAGCAGAGAC[G/T]GAGAGCAGGAACAGAGATGGAGAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP5SL PubMed Links

Gene Details

Gene
ATP5SL
Gene Name
ATP5S like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167867.1 Intron NP_001161339.1
NM_001167868.1 Intron NP_001161340.1
NM_001167869.1 Intron NP_001161341.1
NM_001167870.1 Intron NP_001161342.1
NM_001167871.1 Intron NP_001161343.1
NM_001320838.1 Intron NP_001307767.1
NM_001320839.1 Intron NP_001307768.1
NM_001320840.1 Intron NP_001307769.1
NM_001320841.1 Intron NP_001307770.1
NM_001320844.1 Intron NP_001307773.1
NM_018035.2 Intron NP_060505.2
XM_011527065.2 Intron XP_011525367.1
Gene
ERICH4
Gene Name
glutamate rich 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130514.1 Intron NP_001123986.1
XM_017026119.1 Intron XP_016881608.1

View Full Product Details