Product Details

SNP ID
rs5753674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31520903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTAGTACCAGCTGCTCAGGAGT[C/T]TAAAGTGGGAGGATCACGTGAGCCT
Phenotype
MIM: 612765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFI1 PubMed Links

Gene Details

Gene
SFI1
Gene Name
SFI1 centrin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007467.2 Intron NP_001007468.1
NM_001258325.1 Intron NP_001245254.1
NM_001258326.1 Intron NP_001245255.1
NM_001258327.1 Intron NP_001245256.1
NM_014775.3 Intron NP_055590.2

View Full Product Details