Product Details

SNP ID
rs10483201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39913326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCAGTTCTAGAGTGGATACCAG[A/G]TATGATCTCAAAATCGTGTTAGACA
Phenotype
MIM: 604518
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRAP2 PubMed Links

Gene Details

Gene
GRAP2
Gene Name
GRB2-related adaptor protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291824.1 Intron NP_001278753.1
NM_001291825.1 Intron NP_001278754.1
NM_001291826.1 Intron NP_001278755.1
NM_001291828.1 Intron NP_001278757.1
NM_004810.3 Intron NP_004801.1
XM_006724376.3 Intron XP_006724439.1

View Full Product Details