Product Details

SNP ID
rs10261346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30293952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTACCTATTTATATGTGAGCTGA[A/G]GAAAAAAAGAAGAATGAAGCCATGC
Phenotype
MIM: 612061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105375218 PubMed Links

Gene Details

Gene
LOC105375218
Gene Name
uncharacterized LOC105375218
There are no transcripts associated with this gene.

Gene
MIR550A1
Gene Name
microRNA 550a-1
There are no transcripts associated with this gene.

Gene
MIR550B1
Gene Name
microRNA 550b-1
There are no transcripts associated with this gene.

Gene
ZNRF2
Gene Name
zinc and ring finger 2, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147128.3 Intron NP_667339.1

View Full Product Details