Product Details

SNP ID
rs6435438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208273119 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTTTGTACTGGGAACTGTTTT[A/G]ATAGTGTAAGGGATCTGTCCAGTAA
Phenotype
MIM: 609414
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIKFYVE PubMed Links

Gene Details

Gene
PIKFYVE
Gene Name
phosphoinositide kinase, FYVE-type zinc finger containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178000.1 Intron NP_001171471.1
NM_015040.3 Intron NP_055855.2
NM_152671.3 Intron NP_689884.1
XM_011510778.2 Intron XP_011509080.1
XM_011510779.2 Intron XP_011509081.1
XM_011510780.2 Intron XP_011509082.1
XM_011510781.2 Intron XP_011509083.1
XM_011510782.2 Intron XP_011509084.1
XM_011510783.2 Intron XP_011509085.1
XM_011510784.2 Intron XP_011509086.1
XM_011510785.2 Intron XP_011509087.1
XM_011510786.2 Intron XP_011509088.1
XM_011510787.1 Intron XP_011509089.1
XM_011510788.1 Intron XP_011509090.1
XM_011510789.2 Intron XP_011509091.1
XM_011510790.1 Intron XP_011509092.1
XM_011510792.2 Intron XP_011509094.1
XM_017003568.1 Intron XP_016859057.1
XM_017003569.1 Intron XP_016859058.1
XM_017003570.1 Intron XP_016859059.1
XM_017003571.1 Intron XP_016859060.1
XM_017003572.1 Intron XP_016859061.1
XM_017003573.1 Intron XP_016859062.1
XM_017003574.1 Intron XP_016859063.1

View Full Product Details