Product Details

SNP ID
rs5960531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55908254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGGGGCGGGGAGGCCGCCACTA[A/G]CAAACAACCTCAGCTTCATAAATCC
Phenotype
MIM: 300286
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLF8 PubMed Links

Gene Details

Gene
KLF8
Gene Name
Kruppel like factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159296.2 132 Intron NP_001152768.1
NM_001324099.1 132 Intron NP_001311028.1
NM_001324100.1 132 Intron NP_001311029.1
NM_001324102.1 132 Intron NP_001311031.1
NM_001324104.1 132 Intron NP_001311033.1
NM_001324105.1 132 UTR 5 NP_001311034.1
NM_007250.5 132 Intron NP_009181.2

View Full Product Details