Product Details

SNP ID
rs6103841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44717344 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAATAGCCTCTCCTGGGTATCGG[A/G]GCCTACAGAGTCCTGCCCCCTGACA
Phenotype
MIM: 603399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNK15-AS1 PubMed Links

Gene Details

Gene
KCNK15-AS1
Gene Name
KCNK15 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
WISP2
Gene Name
WNT1 inducible signaling pathway protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323369.1 Intron NP_001310298.1
NM_001323370.1 Intron NP_001310299.1
NM_003881.3 Intron NP_003872.1
XM_017028116.1 Intron XP_016883605.1
XM_017028117.1 Intron XP_016883606.1

View Full Product Details