Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001284251.1 | Intron | NP_001271180.1 | |||
NM_001284252.1 | Intron | NP_001271181.1 | |||
NM_001284253.1 | Intron | NP_001271182.1 | |||
NM_001284254.1 | Intron | NP_001271183.1 | |||
NM_001284255.1 | Intron | NP_001271184.1 | |||
NM_001284256.1 | Intron | NP_001271185.1 | |||
NM_001284257.1 | Intron | NP_001271186.1 | |||
NM_031444.3 | Intron | NP_113632.2 | |||
XM_005261761.1 | Intron | XP_005261818.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278656.1 | Intron | NP_001265585.1 | |||
NM_004175.4 | Intron | NP_004166.1 |