Product Details

SNP ID
rs10232537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149547799 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTGTAACAAGTGAGGGGTGCAA[C/T]TGAAGGTACAGCATTTGCCTGCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF767P PubMed Links

Gene Details

Gene
ZNF767P
Gene Name
zinc finger family member 767, pseudogene
There are no transcripts associated with this gene.

View Full Product Details