Product Details

SNP ID
rs6142309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35172199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATACTGCTGCTGTCTGGCTGGGC[C/T]TTTTGTAGCCAAGACGCCTCAGATG
Phenotype
MIM: 600646
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PROCR PubMed Links

Gene Details

Gene
PROCR
Gene Name
protein C receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006404.4 263 Silent Mutation GCC,GCT A15A NP_006395.2
XM_011528496.1 263 Silent Mutation GCC,GCT A15A XP_011526798.1

View Full Product Details