Product Details

SNP ID
rs6058616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32278613 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTCTGCTCAGAGCAAGGGAGGT[A/G]CTTCTACAAGTAGTCCTGGGAAACT
Phenotype
MIM: 603754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF3B PubMed Links

Gene Details

Gene
KIF3B
Gene Name
kinesin family member 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004798.3 Intron NP_004789.1

View Full Product Details