Product Details

SNP ID
rs132630301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134377639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCATGTCAAGCTCAGTTGAACAG[A/T]AAAAAGGGCCTACAAGACAGCGCAA
Phenotype
MIM: 300414
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PHF6 PubMed Links

Gene Details

Gene
PHF6
Gene Name
PHD finger protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015877.1 224 Nonsense Mutation AAA,TAA K8* NP_001015877.1
NM_032335.3 224 Nonsense Mutation AAA,TAA K8* NP_115711.2
NM_032458.2 224 Nonsense Mutation AAA,TAA K8* NP_115834.1

View Full Product Details