Product Details

SNP ID
rs9423297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122980189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAAACTGAGGCGCCAATCCTAC[C/T]GGCTGGCAACTTTCGGCAGGGAGGG
Phenotype
MIM: 611310
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSTK PubMed Links

Gene Details

Gene
PSTK
Gene Name
phosphoseryl-tRNA kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153336.2 150 UTR 5 NP_699167.2
XM_017015640.1 150 Intron XP_016871129.1
XM_017015641.1 150 Intron XP_016871130.1

View Full Product Details