Product Details

SNP ID
rs6094510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46901709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTGGGAAGGGAGCGGAGAGGAA[A/G]GAGCCCCGAGGAGAATCGTGCACCT
Phenotype
MIM: 601654
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EYA2 PubMed Links

Gene Details

Gene
EYA2
Gene Name
EYA transcriptional coactivator and phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005244.4 Intron NP_005235.3
NM_172110.3 Intron NP_742108.2

View Full Product Details