Product Details

SNP ID
rs6076557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3822319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACCACACAGCCCAGCCCCAGGC[C/T]TTCATTGAACACGTACCTGATTGTA
Phenotype
MIM: 614824
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP5S1 PubMed Links

Gene Details

Gene
AP5S1
Gene Name
adaptor related protein complex 5 sigma 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204446.1 Intron NP_001191375.1
NM_001204447.1 Intron NP_001191376.1
NM_018347.2 Intron NP_060817.1
Gene
LOC101929125
Gene Name
uncharacterized LOC101929125
There are no transcripts associated with this gene.

View Full Product Details