Product Details

SNP ID
rs7536593
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:228651850 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGCTTCAGGAAATAAGCTGATCA[C/T]CATCTAGTCAGGAAAGTAACCCTAC
Phenotype
MIM: 606366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP5P1 PubMed Links

Gene Details

Gene
DUSP5P1
Gene Name
dual specificity phosphatase 5 pseudogene 1
There are no transcripts associated with this gene.

Gene
LOC105373132
Gene Name
uncharacterized LOC105373132
There are no transcripts associated with this gene.

Gene
RHOU
Gene Name
ras homolog family member U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021205.5 Intron NP_067028.1
Gene
RNA5S16
Gene Name
RNA, 5S ribosomal 16
There are no transcripts associated with this gene.

Gene
RNA5S17
Gene Name
RNA, 5S ribosomal 17
There are no transcripts associated with this gene.

View Full Product Details