Product Details

SNP ID
rs10425730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37218971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGCACTCTGGTTCTGAATGGTG[C/G]TATGTTTGAGGGGTCTGTAAATATG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF383 PubMed Links

Gene Details

Gene
ZNF383
Gene Name
zinc finger protein 383
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152604.1 424 Intron NP_689817.1
XM_005258585.2 424 Intron XP_005258642.1
XM_005258587.2 424 Intron XP_005258644.1
XM_005258588.2 424 Intron XP_005258645.1
XM_011526586.2 424 UTR 5 XP_011524888.1
XM_011526587.2 424 Intron XP_011524889.1
XM_011526588.2 424 Intron XP_011524890.1
XM_011526589.2 424 Intron XP_011524891.1
XM_011526590.2 424 Intron XP_011524892.1
XM_017026422.1 424 Intron XP_016881911.1
XM_017026423.1 424 Intron XP_016881912.1
XM_017026424.1 424 Intron XP_016881913.1
Gene
ZNF585B
Gene Name
zinc finger protein 585B
There are no transcripts associated with this gene.

View Full Product Details