Product Details

SNP ID
rs9486269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:106047778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGTTGAGAACCTTTGATAGTTG[A/T]CAGTTTTTACTATGGTTAGCCAAGT
Phenotype
MIM: 603423
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRDM1 PubMed Links

Gene Details

Gene
PRDM1
Gene Name
PR domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198.3 Intron NP_001189.2
NM_182907.2 Intron NP_878911.1
XM_006715550.3 Intron XP_006715613.1
XM_011536062.2 Intron XP_011534364.1
XM_011536063.2 Intron XP_011534365.1
XM_011536064.2 Intron XP_011534366.1
XM_017011187.1 Intron XP_016866676.1

View Full Product Details