Product Details

SNP ID
rs5938009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:75276576 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAACAAAGAAAAATAGATGATGAT[A/T]CTTGTTTCTATAGAGCAGAAGTTAC
Phenotype
MIM: 300656
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UPRT PubMed Links

Gene Details

Gene
UPRT
Gene Name
uracil phosphoribosyltransferase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307944.1 Intron NP_001294873.1
NM_145052.3 Intron NP_659489.1
XM_011530867.2 Intron XP_011529169.1

View Full Product Details