Product Details

SNP ID
rs5967245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101108503 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTCACCACGCTCGGCCCCAAATT[C/T]AGCATTTTTTTAAAGCATAGTCACA
Phenotype
MIM: 300065
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPI PubMed Links

Gene Details

Gene
CENPI
Gene Name
centromere protein I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318521.1 Intron NP_001305450.1
NM_001318523.1 Intron NP_001305452.1
NM_006733.3 Intron NP_006724.2
XM_005262111.2 Intron XP_005262168.1
XM_011530894.2 Intron XP_011529196.1
XM_011530895.2 Intron XP_011529197.1
XM_011530897.2 Intron XP_011529199.1
XM_011530899.2 Intron XP_011529201.1
XM_017029383.1 Intron XP_016884872.1

View Full Product Details