Product Details

SNP ID
rs7530017
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:46559154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCCTCTCCCAGCCTGCGTGGGG[A/T]AAAAGCTCCCAGAGCAAGGCCTGCG
Phenotype
MIM: 611455 MIM: 606724
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KNCN PubMed Links

Gene Details

Gene
KNCN
Gene Name
kinocilin
There are no transcripts associated with this gene.

Gene
MKNK1
Gene Name
MAP kinase interacting serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135553.2 Intron NP_001129025.1
NM_003684.5 Intron NP_003675.2
NM_198973.3 Intron NP_945324.1
XM_006711000.1 Intron XP_006711063.1
XM_006711001.2 Intron XP_006711064.1
XM_006711002.2 Intron XP_006711065.1
XM_006711003.1 Intron XP_006711066.1
XM_006711006.3 Intron XP_006711069.1
XM_006711007.1 Intron XP_006711070.1
XM_006711009.3 Intron XP_006711072.1
XM_006711013.1 Intron XP_006711076.1
XM_006711014.1 Intron XP_006711077.1
XM_006711015.1 Intron XP_006711078.1
XM_017002652.1 Intron XP_016858141.1
XM_017002653.1 Intron XP_016858142.1
XM_017002654.1 Intron XP_016858143.1
XM_017002655.1 Intron XP_016858144.1
XM_017002656.1 Intron XP_016858145.1
XM_017002657.1 Intron XP_016858146.1
XM_017002658.1 Intron XP_016858147.1
XM_017002659.1 Intron XP_016858148.1
XM_017002660.1 Intron XP_016858149.1
XM_017002661.1 Intron XP_016858150.1
XM_017002662.1 Intron XP_016858151.1
XM_017002663.1 Intron XP_016858152.1
XM_017002664.1 Intron XP_016858153.1
XM_017002665.1 Intron XP_016858154.1
XM_017002666.1 Intron XP_016858155.1
XM_017002667.1 Intron XP_016858156.1
XM_017002668.1 Intron XP_016858157.1
Gene
MKNK1-AS1
Gene Name
MKNK1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details