Product Details

SNP ID
rs9294868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:70415806 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAATTTCTAGTCAGAATATATGA[A/G]CTCTTATGATGACTCATGACATTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM135A PubMed Links

Gene Details

Gene
FAM135A
Gene Name
family with sequence similarity 135 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105531.2 Intron NP_001099001.1
NM_001162529.1 Intron NP_001156001.1
NM_020819.4 Intron NP_065870.3
XM_005248743.1 Intron XP_005248800.1
XM_005248744.4 Intron XP_005248801.1
XM_005248745.2 Intron XP_005248802.1
XM_005248748.2 Intron XP_005248805.1
XM_005248749.1 Intron XP_005248806.1
XM_005248751.2 Intron XP_005248808.1
XM_011535993.2 Intron XP_011534295.1
XM_011535994.2 Intron XP_011534296.1
XM_011535995.2 Intron XP_011534297.1
XM_011535996.2 Intron XP_011534298.1
XM_011535997.2 Intron XP_011534299.1
XM_017011124.1 Intron XP_016866613.1
XM_017011125.1 Intron XP_016866614.1
XM_017011126.1 Intron XP_016866615.1
XM_017011127.1 Intron XP_016866616.1
XM_017011128.1 Intron XP_016866617.1
XM_017011129.1 Intron XP_016866618.1
XM_017011130.1 Intron XP_016866619.1
XM_017011131.1 Intron XP_016866620.1
XM_017011132.1 Intron XP_016866621.1
XM_017011133.1 Intron XP_016866622.1
XM_017011134.1 Intron XP_016866623.1
XM_017011135.1 Intron XP_016866624.1
XM_017011136.1 Intron XP_016866625.1

View Full Product Details