Product Details

SNP ID
rs6060017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34725238 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAGGCACCAGAGATACAGTAGTT[A/C]GCAAAATCAGACATAATCTCCTGCC
Phenotype
MIM: 605299
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NCOA6 PubMed Links

Gene Details

Gene
NCOA6
Gene Name
nuclear receptor coactivator 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242539.2 Intron NP_001229468.1
NM_001318240.1 Intron NP_001305169.1
NM_014071.4 Intron NP_054790.2
XM_006723755.3 Intron XP_006723818.1
XM_011528721.2 Intron XP_011527023.1
XM_011528726.2 Intron XP_011527028.1
XM_011528727.2 Intron XP_011527029.1
XM_017027739.1 Intron XP_016883228.1
XM_017027740.1 Intron XP_016883229.1
XM_017027741.1 Intron XP_016883230.1
XM_017027742.1 Intron XP_016883231.1
XM_017027743.1 Intron XP_016883232.1
XM_017027744.1 Intron XP_016883233.1
XM_017027745.1 Intron XP_016883234.1
XM_017027746.1 Intron XP_016883235.1
XM_017027747.1 Intron XP_016883236.1
XM_017027748.1 Intron XP_016883237.1
XM_017027749.1 Intron XP_016883238.1
XM_017027750.1 Intron XP_016883239.1
XM_017027751.1 Intron XP_016883240.1

View Full Product Details