Product Details

SNP ID
rs8111157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11147962 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCCAGCACCAACTTCAACCAAGA[A/G]CCGGACTGCACAAGGAGGCACAGGC
Phenotype
MIM: 609394
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPC24 PubMed Links

Gene Details

Gene
SPC24
Gene Name
SPC24, NDC80 kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317031.1 Intron NP_001303960.1
NM_001317032.1 Intron NP_001303961.1
NM_001317033.1 Intron NP_001303962.1
NM_182513.3 Intron NP_872319.1
XM_005259753.3 Intron XP_005259810.1
XM_011527702.1 Intron XP_011526004.1

View Full Product Details