Product Details
- SNP ID
-
rs10016763
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:44681678 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCGTAAAGACACTCAGCAGATGA[A/C]CATGTAAGAGCTGTGAATATAAACT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GUF1
PubMed Links
Gene Details
- Gene
- GUF1
- Gene Name
- GUF1 homolog, GTPase
View Full Product Details