Product Details

SNP ID
rs9934438
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.16:31093557 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
CCCCGACCTCCCATCCTAGTCCAAG[A/G]GTCGATGATCTCCTGGCACCGGGCA
Phenotype
MIM: 610561 MIM: 608547
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
There are no transcripts associated with this gene.

Gene
VKORC1
Gene Name
vitamin K epoxide reductase complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311311.1 Intron NP_001298240.1
NM_024006.5 Intron NP_076869.1
NM_206824.2 Intron NP_996560.1

View Full Product Details