Product Details

SNP ID
rs5962953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104576966 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTACATATTCAGCTTTCTTTCC[C/T]ATCTGTTTCCCATGGTAGTTATTTC
Phenotype
MIM: 300277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL1RAPL2 PubMed Links

Gene Details

Gene
IL1RAPL2
Gene Name
interleukin 1 receptor accessory protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017416.1 Intron NP_059112.1
XM_011530905.2 Intron XP_011529207.1
XM_017029403.1 Intron XP_016884892.1

View Full Product Details