Product Details

SNP ID
rs6021696
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:52085949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACACAAAGATAAGCATCTTATC[C/T]TACACACCCATCCTTTCCCTAACCT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZFP64 PubMed Links

Gene Details

Gene
ZFP64
Gene Name
ZFP64 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319146.1 Intron NP_001306075.1
NM_018197.2 Intron NP_060667.2
NM_022088.4 Intron NP_071371.3
NM_199426.1 Intron NP_955458.1
NM_199427.2 Intron NP_955459.2

View Full Product Details